Product Details

SNP ID
rs201996792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:39460804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTTCTTTAGGTATTTGGGAGATA[C/T]TTTTGCAGCCCAGTCAGACCGTTAA
Phenotype
MIM: 611135 MIM: 607031 MIM: 603686
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLB PubMed Links

Gene Details

Gene
KLB
Gene Name
klotho beta
There are no transcripts associated with this gene.

Gene
LIAS
Gene Name
lipoic acid synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278590.1 154 Silent Mutation TAC,TAT Y20Y NP_001265519.1
NM_001278591.1 154 Silent Mutation TAC,TAT Y20Y NP_001265520.1
NM_001278592.1 154 Silent Mutation TAC,TAT Y20Y NP_001265521.1
NM_006859.3 154 Silent Mutation TAC,TAT Y20Y NP_006850.2
NM_194451.2 154 Silent Mutation TAC,TAT Y20Y NP_919433.1
XM_006713990.3 154 Silent Mutation TAC,TAT Y20Y XP_006714053.1
XM_017007665.1 154 Silent Mutation TAC,TAT Y20Y XP_016863154.1
XM_017007666.1 154 Silent Mutation TAC,TAT Y20Y XP_016863155.1
XM_017007667.1 154 Silent Mutation TAC,TAT Y20Y XP_016863156.1
Gene
RPL9
Gene Name
ribosomal protein L9
There are no transcripts associated with this gene.

View Full Product Details