Product Details

SNP ID
rs201668488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:73061849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAGCAATTGGTATCATCAACAC[C/T]GACATTGCACGGACAAAGTACGTAA
Phenotype
MIM: 610428
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COX18 PubMed Links

Gene Details

Gene
COX18
Gene Name
COX18, cytochrome c oxidase assembly factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297732.1 905 Silent Mutation NP_001284661.1
NM_001297733.1 905 Silent Mutation NP_001284662.1
NM_001300729.1 905 Silent Mutation NP_001287658.1
NM_173827.3 905 Silent Mutation NP_776188.1
XM_005265680.4 905 Silent Mutation XP_005265737.1
XM_011531878.2 905 Silent Mutation XP_011530180.1
XM_017008045.1 905 Silent Mutation XP_016863534.1

View Full Product Details