Product Details

SNP ID
rs201187038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:4387754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCAGCTGCAGTTCCCGCCCCCGG[A/C]TAAGGTAAGCCCCCCCACGCCCCTC
Phenotype
MIM: 607645
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NSG1 PubMed Links

Gene Details

Gene
NSG1
Gene Name
neuron specific gene family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040101.1 362 Missense Mutation GAT,GCT D42A NP_001035190.1
NM_001287763.1 362 Missense Mutation GAT,GCT D42A NP_001274692.1
NM_001287764.1 362 Missense Mutation GAT,GCT D15A NP_001274693.1
NM_014392.4 362 Missense Mutation GAT,GCT D42A NP_055207.1
XM_017008022.1 362 Missense Mutation GAT,GCT D42A XP_016863511.1

View Full Product Details