Product Details

SNP ID
rs201674202
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:57033267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATGGCCAGGTTGTCCCGGTCACC[A/G]GGCAGGAGTTCTGTCCTTTGAACTC
Phenotype
MIM: 602867 MIM: 180661
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
IGFBP7 PubMed Links

Gene Details

Gene
IGFBP7
Gene Name
insulin like growth factor binding protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253835.1 664 Silent Mutation CCC,CCT P210P NP_001240764.1
NM_001553.2 664 Silent Mutation CCC,CCT P210P NP_001544.1
Gene
POLR2B
Gene Name
RNA polymerase II subunit B
There are no transcripts associated with this gene.

View Full Product Details