Product Details

SNP ID
rs201248791
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:143185533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGATTGTGCGGAAGGTGGTGGAAT[C/T]GGCGGAGCACTGGCTAGACGAGGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105377623 PubMed Links

Gene Details

Gene
LOC105377623
Gene Name
uncharacterized LOC105377623
There are no transcripts associated with this gene.

Gene
USP38
Gene Name
ubiquitin specific peptidase 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290325.1 617 Missense Mutation TCG,TTG S28L NP_001277254.1
NM_001290326.1 617 UTR 5 NP_001277255.1
NM_032557.6 617 Missense Mutation TCG,TTG S28L NP_115946.2
XM_011532360.2 617 Missense Mutation TCG,TTG S28L XP_011530662.1

View Full Product Details