Product Details
- SNP ID
-
hCV191443117
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:42762546 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAGCAAAGGAGGTCTCCATTAAAG[A/G]AGGTTAGTTATTTGCTGCCTGAGGA
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC152
PubMed Links
Gene Details
- Gene
- CCDC152
- Gene Name
- coiled-coil domain containing 152
There are no transcripts associated with this gene.
View Full Product Details