Product Details

SNP ID
rs202025173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138508718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACTGAGACCCTTCTTGTGATTT[A/G]TCTGTGACAATTTCCAACGTAGCTC
Phenotype
MIM: 600285
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ETF1 PubMed Links

Gene Details

Gene
ETF1
Gene Name
eukaryotic translation termination factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256302.1 1603 Silent Mutation GAC,GAT D361D NP_001243231.1
NM_001282185.1 1603 Silent Mutation GAC,GAT D380D NP_001269114.1
NM_001291974.1 1603 Silent Mutation GAC,GAT D361D NP_001278903.1
NM_001291975.1 1603 Silent Mutation GAC,GAT D361D NP_001278904.1
NM_004730.3 1603 Silent Mutation GAC,GAT D394D NP_004721.1
XM_005271921.1 1603 Silent Mutation GAC,GAT D361D XP_005271978.1
XM_017009221.1 1603 Silent Mutation GAC,GAT D342D XP_016864710.1
XM_017009222.1 1603 Silent Mutation GAC,GAT D179D XP_016864711.1

View Full Product Details