Product Details

SNP ID
rs202241708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:78998117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTTTCCACCTTTTTTCTGAAGCC[A/G]GTTTCTGGTTGGTTCGGTCAATACC
Phenotype
MIM: 605849
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DMGDH PubMed Links

Gene Details

Gene
DMGDH
Gene Name
dimethylglycine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013391.3 2620 Missense Mutation CGG,TGG R856W NP_037523.2
XM_006714597.2 2620 Intron XP_006714660.1
XM_011543354.2 2620 Intron XP_011541656.1
XM_011543355.2 2620 Intron XP_011541657.1

View Full Product Details