Product Details

SNP ID
rs202048719
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141339512 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCCAAATGACCACTTCTCCCTG[G/T]ACGTGCGAAGGGGAGCTGATGGGAA
Phenotype
MIM: 606288 MIM: 606289 MIM: 606290
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDHGA1 PubMed Links

Gene Details

Gene
PCDHGA1
Gene Name
protocadherin gamma subfamily A, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018912.2 753 Intron NP_061735.1
NM_031993.1 753 Intron NP_114382.1
Gene
PCDHGA2
Gene Name
protocadherin gamma subfamily A, 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018915.3 753 Missense Mutation GAC,TAC D181Y NP_061738.1
NM_032009.2 753 Missense Mutation GAC,TAC D181Y NP_114398.1
Gene
PCDHGA3
Gene Name
protocadherin gamma subfamily A, 3
There are no transcripts associated with this gene.

View Full Product Details