Product Details

SNP ID
rs201959000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141403384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTCTGGAAGTAAAAATTAACGA[A/C]ATCGCGGTTCCTGGAGCACGTTATC
Phenotype
MIM: 606288 MIM: 606297 MIM: 606289 MIM: 606290 MIM: 606291 MIM: 606292 MIM: 606293 MIM: 606294 MIM: 606295 MIM: 606296 MIM: 606299 MIM: 606300 MIM: 606301 MIM: 603058 MIM: 606302 MIM: 606303
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PCDHGA1 PubMed Links

Gene Details

Gene
PCDHGA1
Gene Name
protocadherin gamma subfamily A, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018912.2 432 Intron NP_061735.1
NM_031993.1 432 Intron NP_114382.1
Gene
PCDHGA10
Gene Name
protocadherin gamma subfamily A, 10
There are no transcripts associated with this gene.

Gene
PCDHGA2
Gene Name
protocadherin gamma subfamily A, 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018915.3 432 Intron NP_061738.1
NM_032009.2 432 Intron NP_114398.1
Gene
PCDHGA3
Gene Name
protocadherin gamma subfamily A, 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018916.3 432 Intron NP_061739.2
NM_032011.1 432 Intron NP_114400.1
Gene
PCDHGA4
Gene Name
protocadherin gamma subfamily A, 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018917.3 432 Intron NP_061740.2
NM_032053.2 432 Intron NP_114442.2
Gene
PCDHGA5
Gene Name
protocadherin gamma subfamily A, 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018918.2 432 Intron NP_061741.1
NM_032054.1 432 Intron NP_114443.1
Gene
PCDHGA6
Gene Name
protocadherin gamma subfamily A, 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018919.2 432 Intron NP_061742.1
NM_032086.1 432 Intron NP_114475.1
Gene
PCDHGA7
Gene Name
protocadherin gamma subfamily A, 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018920.3 432 Intron NP_061743.1
NM_032087.2 432 Intron NP_114476.1
Gene
PCDHGA8
Gene Name
protocadherin gamma subfamily A, 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014004.2 432 Intron NP_054723.1
NM_032088.1 432 Intron NP_114477.1
Gene
PCDHGA9
Gene Name
protocadherin gamma subfamily A, 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018921.2 432 Missense Mutation GAA,GAC E144D NP_061744.1
NM_032089.1 432 Missense Mutation GAA,GAC E144D NP_114478.1
Gene
PCDHGB1
Gene Name
protocadherin gamma subfamily B, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018922.2 432 Intron NP_061745.1
NM_032095.1 432 Intron NP_115266.1
Gene
PCDHGB2
Gene Name
protocadherin gamma subfamily B, 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018923.2 432 Intron NP_061746.1
NM_032096.1 432 Intron NP_115267.1
Gene
PCDHGB3
Gene Name
protocadherin gamma subfamily B, 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018924.4 432 Intron NP_061747.2
NM_032097.2 432 Intron NP_115268.2
Gene
PCDHGB4
Gene Name
protocadherin gamma subfamily B, 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003736.2 432 Intron NP_003727.1
NM_032098.1 432 Intron NP_115269.1
Gene
PCDHGB5
Gene Name
protocadherin gamma subfamily B, 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018925.2 432 Intron NP_061748.1
NM_032099.1 432 Intron NP_115270.1
Gene
PCDHGB6
Gene Name
protocadherin gamma subfamily B, 6
There are no transcripts associated with this gene.

View Full Product Details