Product Details
- SNP ID
-
rs201900739
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:179678675 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATCTTCCTCTGCCCGGCGCGCGCCG[C/T]AGCCGTCGGGATCACCTCGGGGTTG
- Phenotype
-
MIM: 114217
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CANX
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78613293] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CANX
- Gene Name
- calnexin
- Gene
- CBY3
- Gene Name
- chibby family member 3
- Gene
- LOC105377763
- Gene Name
- uncharacterized LOC105377763
There are no transcripts associated with this gene.
View Full Product Details