Product Details

SNP ID
rs200341894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139478434 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGTCCAATGGGAGGAGAATAT[A/C]CAGCCGCTGGCTCACTGCACCCCGT
Phenotype
MIM: 612374
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM173 PubMed Links

Gene Details

Gene
TMEM173
Gene Name
transmembrane protein 173
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301738.1 859 Missense Mutation GAT,TAT D199Y NP_001288667.1
NM_198282.3 859 Missense Mutation GAT,TAT D199Y NP_938023.1
XM_005268445.3 859 Missense Mutation GAT,TAT D199Y XP_005268502.1
XM_011537639.2 859 Missense Mutation GAT,TAT D199Y XP_011535941.1
XM_011537640.2 859 Missense Mutation GAT,TAT D80Y XP_011535942.1

View Full Product Details