Product Details
- SNP ID
-
rs199985535
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:155014116 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGGATATAATGCAACGGTCCTGG[A/C]CTATGGGCAGACTGGCTCTGGAAAA
- Phenotype
-
MIM: 609184
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KIF4B
PubMed Links
Gene Details
- Gene
- KIF4B
- Gene Name
- kinesin family member 4B
There are no transcripts associated with this gene.
View Full Product Details