Product Details

SNP ID
rs200285354
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:137753974 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCACCACGCAGTCCGTCAGAGT[C/T]CCAAAGGCCTCAAAGTGGCCGCGCA
Phenotype
MIM: 609409
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HNRNPA0 PubMed Links

Gene Details

Gene
HNRNPA0
Gene Name
heterogeneous nuclear ribonucleoprotein A0
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006805.3 377 Silent Mutation GGA,GGG G31G NP_006796.1

View Full Product Details