Product Details

SNP ID
rs199593005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43492267 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAATTAACTGCTCTTGTCCATCAG[A/G]AAAAGTTAACTTACACCAACCTAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C5orf34 PubMed Links

Gene Details

Gene
C5orf34
Gene Name
chromosome 5 open reading frame 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198566.3 1871 Missense Mutation CCT,TCT P510S NP_940968.1
XM_011514038.2 1871 Missense Mutation CCT,TCT P510S XP_011512340.1
XM_017009445.1 1871 Missense Mutation CCT,TCT P396S XP_016864934.1
XM_017009446.1 1871 Missense Mutation CCT,TCT P396S XP_016864935.1
Gene
TMEM267
Gene Name
transmembrane protein 267
There are no transcripts associated with this gene.

View Full Product Details