Product Details

SNP ID
rs200897371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43039860 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGCCAGTAGACTCCGGGCAGCC[A/G]CCAGCAAGGGCTGGAAAGCAGTCCC
Phenotype
MIM: 611296
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANXA2R PubMed Links

Gene Details

Gene
ANXA2R
Gene Name
annexin A2 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014279.2 486 Missense Mutation CGG,TGG R63W NP_001014301.1
Gene
LOC153684
Gene Name
uncharacterized LOC153684
There are no transcripts associated with this gene.

Gene
LOC648987
Gene Name
uncharacterized LOC648987
There are no transcripts associated with this gene.

View Full Product Details