Product Details

SNP ID
rs200453473
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154891257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACTTACCACTGGCTCTGAGAAC[G/T]GCAGAGGGGCTGCTCTGCTTCTACT
Phenotype
MIM: 607005
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GEMIN5 PubMed Links

Gene Details

Gene
GEMIN5
Gene Name
gem nuclear organelle associated protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252156.1 4326 Missense Mutation AGT,CGT S1415R NP_001239085.1
NM_015465.4 4326 Missense Mutation AGT,CGT S1416R NP_056280.2

View Full Product Details