Product Details

SNP ID
rs201508591
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:145859736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGACATGCAAAAGCCACGGGCTA[C/T]TGATTGCAAATCTGGCAAGGCTGTA
Phenotype
MIM: 615762
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRXCR2 PubMed Links

Gene Details

Gene
GRXCR2
Gene Name
glutaredoxin and cysteine rich domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080516.1 744 Silent Mutation CAA,CAG Q248Q NP_001073985.1
XM_017009708.1 744 Silent Mutation CAA,CAG Q152Q XP_016865197.1

View Full Product Details