Product Details

SNP ID
rs201055359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141945692 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGTCTGGAAGGTCCTTGGCTCCT[C/G]CGTGGCTGCAGCATCCGGGGAGATC
Phenotype
MIM: 615741 MIM: 605622 MIM: 605675
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0141 PubMed Links

Gene Details

Gene
KIAA0141
Gene Name
KIAA0141
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142603.2 4455 Intron NP_001136075.1
NM_014773.4 4455 Intron NP_055588.3
Gene
PCDH12
Gene Name
protocadherin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016580.3 4455 Missense Mutation CAG,GAG Q1082E NP_057664.1
Gene
RNF14
Gene Name
ring finger protein 14
There are no transcripts associated with this gene.

View Full Product Details