Product Details

SNP ID
rs202165984
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33263504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACCTGTGTCCGAGCATTGAGCA[A/G]CTGCTGGAAGCTCTCAACCTCTTTG
Phenotype
MIM: 180473 MIM: 603443
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HCG25 PubMed Links

Gene Details

Gene
HCG25
Gene Name
HLA complex group 25 (non-protein coding)
There are no transcripts associated with this gene.

Gene
RPS18
Gene Name
ribosomal protein S18
There are no transcripts associated with this gene.

Gene
VPS52
Gene Name
VPS52, GARP complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289174.1 1999 Silent Mutation CTG,TTG L525L NP_001276103.1
NM_001289175.1 1999 Silent Mutation CTG,TTG L467L NP_001276104.1
NM_001289176.1 1999 Silent Mutation CTG,TTG L403L NP_001276105.1
NM_022553.5 1999 Silent Mutation CTG,TTG L592L NP_072047.4
XM_011514797.1 1999 Silent Mutation CTG,TTG L525L XP_011513099.1
XM_011514798.2 1999 Silent Mutation CTG,TTG L525L XP_011513100.1
XM_011514799.1 1999 Silent Mutation CTG,TTG L525L XP_011513101.1
XM_017011177.1 1999 Silent Mutation CTG,TTG L592L XP_016866666.1
XM_017011178.1 1999 Silent Mutation CTG,TTG L467L XP_016866667.1
XM_017011179.1 1999 Intron XP_016866668.1

View Full Product Details