Product Details

SNP ID
rs201995170
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:137001775 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACAGCCTGCCAGTTTGGGGATCC[C/T]AGTCGACCAGGGTCGTCGATGGCTC
Phenotype
MIM: 605620
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL20RA PubMed Links

Gene Details

Gene
IL20RA
Gene Name
interleukin 20 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278722.1 1718 Nonsense Mutation TAG,TGG *433W NP_001265651.1
NM_001278723.1 1718 Nonsense Mutation TAG,TGG *371W NP_001265652.1
NM_001278724.1 1718 Nonsense Mutation TAG,TGG *349W NP_001265653.1
NM_014432.3 1718 Nonsense Mutation TAG,TGG *482W NP_055247.3
XM_006715506.2 1718 Nonsense Mutation TAG,TGG *349W XP_006715569.1
XM_011535904.2 1718 Nonsense Mutation TAG,TGG *371W XP_011534206.1
XM_017010954.1 1718 Nonsense Mutation TAG,TGG *483W XP_016866443.1
XM_017010955.1 1718 Nonsense Mutation TAG,TGG *434W XP_016866444.1
Gene
NHEG1
Gene Name
neuroblastoma highly expressed 1
There are no transcripts associated with this gene.

View Full Product Details