Product Details

SNP ID
rs201154224
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52757867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTAACTCAGAACCTACCTCTAC[A/T]CCAGCTGCAGCCAGGAGCCACCGGA
Phenotype
MIM: 138360
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GSTA2 PubMed Links

Gene Details

Gene
GSTA2
Gene Name
glutathione S-transferase alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000846.4 231 Silent Mutation GGA,GGT G27G NP_000837.3
XM_011514532.2 231 Intron XP_011512834.1

View Full Product Details