Product Details

SNP ID
rs200763450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:89257889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTCTGTCAGGATATGGCTTCT[G/T]GGGTACCCAGCCAGGCTGTTGGCCT
Phenotype
MIM: 137162
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GABRR2 PubMed Links

Gene Details

Gene
GABRR2
Gene Name
gamma-aminobutyric acid type A receptor rho2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002043.4 2398 Silent Mutation CCA,CCC P393P NP_002034.3
XM_011535713.2 2398 Silent Mutation CCA,CCC P349P XP_011534015.1
XM_011535714.2 2398 Silent Mutation CCA,CCC P294P XP_011534016.1
XM_011535715.2 2398 Silent Mutation CCA,CCC P294P XP_011534017.1
XM_011535716.2 2398 Silent Mutation CCA,CCC P294P XP_011534018.1
XM_011535717.2 2398 Silent Mutation CCA,CCC P294P XP_011534019.1

View Full Product Details