Product Details

SNP ID
rs200486002
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30562010 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTCCTTTTCCGGGGCCAGGCCAT[G/T]GGGGTCCCACCAGGGGAAGCTTTCA
Phenotype
MIM: 603429 MIM: 143024
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCF1 PubMed Links

Gene Details

Gene
ABCF1
Gene Name
ATP binding cassette subfamily F member 1
There are no transcripts associated with this gene.

Gene
GNL1
Gene Name
G protein nucleolar 1 (putative)
There are no transcripts associated with this gene.

Gene
PRR3
Gene Name
proline rich 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077497.2 919 Missense Mutation GGG,TGG G95W NP_001070965.1
NM_025263.3 919 Missense Mutation GGG,TGG G116W NP_079539.2

View Full Product Details