Product Details

SNP ID
rs201200534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:30578181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCTTAAGAAGCTCTCAGTGCCA[A/G]CCAGTGATGAGGAGGATGAAGGTAA
Phenotype
MIM: 603429 MIM: 611619
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCF1 PubMed Links

Gene Details

Gene
ABCF1
Gene Name
ATP binding cassette subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025091.1 417 Missense Mutation ACC,GCC T108A NP_001020262.1
NM_001090.2 417 Missense Mutation ACC,GCC T108A NP_001081.1
Gene
MIR877
Gene Name
microRNA 877
There are no transcripts associated with this gene.

View Full Product Details