Product Details
- SNP ID
-
rs200848146
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:52752872 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTTACTTTTTCAAAGGCAGGGAA[A/G]TAGCGATTTTTTGTTTTCTCTTGGA
- Phenotype
-
MIM: 138360
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GSTA2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2180314,rs2234951] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GSTA2
- Gene Name
- glutathione S-transferase alpha 2
- Gene
- GSTA7P
- Gene Name
- glutathione S-transferase alpha 7, pseudogene
There are no transcripts associated with this gene.
View Full Product Details