Product Details

SNP ID
rs200513000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31506266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAAAACCTGGAGACTCAAGAAT[C/T]GACAGTGCCCCAGTCCTCCAGAGCT
Phenotype
MIM: 602436
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MICB PubMed Links

Gene Details

Gene
MICB
Gene Name
MHC class I polypeptide-related sequence B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289160.1 494 Missense Mutation TCG,TTG S118L NP_001276089.1
NM_001289161.1 494 Intron NP_001276090.1
NM_005931.4 494 Missense Mutation TCG,TTG S150L NP_005922.2

View Full Product Details