Product Details

SNP ID
rs200536541
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:37171354 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCAGGTGCTGGAGGCCGTGCGGC[C/A]CTGCCACAACTGCGGGGTGCTCCAC
Phenotype
MIM: 164960
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
PIM1 PubMed Links

Gene Details

Gene
PIM1
Gene Name
Pim-1 proto-oncogene, serine/threonine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243186.1 900 Missense Mutation CAC,CCC H248P NP_001230115.1
NM_002648.3 900 Missense Mutation CAC,CCC H157P NP_002639.1

View Full Product Details