Product Details

SNP ID
rs200893299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28506345 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAAGTATTTCTGAGTTTGTTCCT[A/G]GTTCTTGTTTTCCAAACTGGTTGCA
Phenotype
MIM: 607913
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPX6 PubMed Links

Gene Details

Gene
GPX6
Gene Name
glutathione peroxidase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182701.1 376 Missense Mutation NP_874360.1

View Full Product Details