Product Details

SNP ID
rs201509607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31719119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAGTGCAGCAGCCAGAGGCCAA[C/G]GCCAGCCAAGGAGGTAAGGAAGACA
Phenotype
MIM: 606520 MIM: 604744 MIM: 610435 MIM: 606038 MIM: 610437 MIM: 611404
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C6orf25 PubMed Links

Gene Details

Gene
C6orf25
Gene Name
chromosome 6 open reading frame 25
There are no transcripts associated with this gene.

Gene
DDAH2
Gene Name
dimethylarginine dimethylaminohydrolase 2
There are no transcripts associated with this gene.

Gene
LY6G6C
Gene Name
lymphocyte antigen 6 complex, locus G6C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025261.2 410 Missense Mutation CTT,GTT L119V NP_079537.1
Gene
LY6G6D
Gene Name
lymphocyte antigen 6 complex, locus G6D
There are no transcripts associated with this gene.

Gene
LY6G6E
Gene Name
lymphocyte antigen 6 complex, locus G6E (pseudogene)
There are no transcripts associated with this gene.

Gene
LY6G6F
Gene Name
lymphocyte antigen 6 complex, locus G6F
There are no transcripts associated with this gene.

View Full Product Details