Product Details

SNP ID
rs199673000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:32157686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATGCCAGCAGCTTCCTGGCCCT[A/G]ATCAATGGGGAAAGAGACCATCCCA
Phenotype
MIM: 609897 MIM: 603298
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EGFL8 PubMed Links

Gene Details

Gene
EGFL8
Gene Name
EGF like domain multiple 8
There are no transcripts associated with this gene.

Gene
LOC100507547
Gene Name
uncharacterized LOC100507547
There are no transcripts associated with this gene.

Gene
PPT2
Gene Name
palmitoyl-protein thioesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204103.1 995 Silent Mutation CTA,CTG L197L NP_001191032.1
NM_005155.6 995 Silent Mutation CTA,CTG L197L NP_005146.4
NM_138717.2 995 Silent Mutation CTA,CTG L203L NP_619731.2
Gene
PPT2-EGFL8
Gene Name
PPT2-EGFL8 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
PRRT1
Gene Name
proline rich transmembrane protein 1
There are no transcripts associated with this gene.

View Full Product Details