Product Details
- SNP ID
-
rs199692317
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:118465484 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTCCTTAATTCATCAATGTCTCTT[C/T]GTAACTGGCACACATCTGACAGCTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CEP85L
PubMed Links
Gene Details
- Gene
- CEP85L
- Gene Name
- centrosomal protein 85 like
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001042475.2 |
3518 |
Missense Mutation |
CAA,CGA |
Q780R |
NP_001035940.1 |
NM_001178035.1 |
3518 |
Missense Mutation |
CAA,CGA |
Q783R |
NP_001171506.1 |
NM_206921.2 |
3518 |
Intron |
|
|
NP_996804.2 |
XM_005266970.1 |
3518 |
Missense Mutation |
CAA,CGA |
Q678R |
XP_005267027.1 |
XM_005266971.1 |
3518 |
Missense Mutation |
CAA,CGA |
Q678R |
XP_005267028.1 |
XM_005266972.4 |
3518 |
Missense Mutation |
CAA,CGA |
Q418R |
XP_005267029.1 |
XM_006715475.3 |
3518 |
Missense Mutation |
CAA,CGA |
Q678R |
XP_006715538.1 |
XM_011535809.2 |
3518 |
Missense Mutation |
CAA,CGA |
Q780R |
XP_011534111.1 |
XM_011535810.2 |
3518 |
Missense Mutation |
CAA,CGA |
Q723R |
XP_011534112.1 |
XM_011535811.1 |
3518 |
Missense Mutation |
CAA,CGA |
Q678R |
XP_011534113.1 |
XM_017010846.1 |
3518 |
Missense Mutation |
CAA,CGA |
Q783R |
XP_016866335.1 |
XM_017010847.1 |
3518 |
Missense Mutation |
CAA,CGA |
Q720R |
XP_016866336.1 |
View Full Product Details