Product Details

SNP ID
rs199760621
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:152697968 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCTTGCTGTACTACAAAGAGA[A/T]AGAATCAAACTGCTTTTTTTCGACA
Phenotype
MIM: 616805
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MYCT1 PubMed Links

Gene Details

Gene
MYCT1
Gene Name
myc target 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025107.2 74 Missense Mutation GAA,GAT E22D NP_079383.2
XM_011536149.2 74 Missense Mutation GAA,GAT E22D XP_011534451.1
XM_011536150.2 74 Missense Mutation GAA,GAT E22D XP_011534452.1
XM_011536151.2 74 Missense Mutation GAA,GAT E22D XP_011534453.1
XM_011536152.2 74 Missense Mutation GAA,GAT E22D XP_011534454.1
XM_017011319.1 74 Missense Mutation GAA,GAT E22D XP_016866808.1

View Full Product Details