Product Details

SNP ID
rs200284390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:29659393 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATGAAGTGGAATTGCCATGTCGC[A/G]TATCTCCTGGGAAGAACGCTACAGG
Phenotype
MIM: 159465
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MOG PubMed Links

Gene Details

Gene
MOG
Gene Name
myelin oligodendrocyte glycoprotein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008228.2 392 Missense Mutation ATA,GTA I55V NP_001008229.1
NM_001008229.2 392 Missense Mutation ATA,GTA I55V NP_001008230.1
NM_001170418.1 392 Intron NP_001163889.1
NM_002433.4 392 Missense Mutation ATA,GTA I55V NP_002424.3
NM_206809.3 392 Missense Mutation ATA,GTA I55V NP_996532.2
NM_206810.3 392 Missense Mutation ATA,GTA I55V NP_996533.2
NM_206811.3 392 Missense Mutation ATA,GTA I55V NP_996534.2
NM_206812.3 392 Missense Mutation ATA,GTA I55V NP_996535.2
NM_206814.5 392 Intron NP_996537.3
XM_005249131.3 392 Missense Mutation ATA,GTA I55V XP_005249188.1

View Full Product Details