Product Details

SNP ID
rs200291392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43623772 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTCCAACAACAGTCCCCACCTC[C/T]GGTACTGTGTAGATTTCATCCACCT
Phenotype
MIM: 607434 MIM: 603968
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GTPBP2 PubMed Links

Gene Details

Gene
GTPBP2
Gene Name
GTP binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286216.1 1344 Silent Mutation CCA,CCG P332P NP_001273145.1
NM_019096.4 1344 Silent Mutation CCA,CCG P420P NP_061969.3
XM_017010976.1 1344 Silent Mutation CCA,CCG P420P XP_016866465.1
Gene
MAD2L1BP
Gene Name
MAD2L1 binding protein
There are no transcripts associated with this gene.

Gene
POLH
Gene Name
DNA polymerase eta
There are no transcripts associated with this gene.

View Full Product Details