Product Details
- SNP ID
-
rs199808617
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:138092096 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCATTGCCCAGAAGGTCATCTTC[A/G]TAGTTGGGGAGGCAGCAGAAGAAGA
- Phenotype
-
MIM: 609301
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PERP
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs648396] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PERP
- Gene Name
- PERP, TP53 apoptosis effector
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022121.4 |
711 |
Silent Mutation |
TAC,TAT |
Y176Y |
NP_071404.2 |
View Full Product Details