Product Details

SNP ID
rs200737615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:121329751 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACACGAGAGATGGAACTGCATG[A/T]TCACCGCCGCCGCCACTACCGCCCC
Phenotype
MIM: 606267
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
WNT16 PubMed Links

Gene Details

Gene
WNT16
Gene Name
Wnt family member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016087.2 537 Missense Mutation ATC,TTC I84F NP_057171.2
NM_057168.1 537 Missense Mutation ATC,TTC I94F NP_476509.1

View Full Product Details