Product Details
- SNP ID
-
rs200869206
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:149070665 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGTCACACTCAGGGCAGTGAAAAG[C/G]CCTCTCCCCTGTGTGCAGGCCCTGA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF786
PubMed Links
Gene Details
- Gene
- ZNF786
- Gene Name
- zinc finger protein 786
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152411.3 |
2244 |
Missense Mutation |
CCT,GCT |
P703A |
NP_689624.2 |
View Full Product Details