Product Details
- SNP ID
-
hCV191748886
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:73865689 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTGGCACCTGGCCTATCTCATCA[C/T]CTGGACCACCTGCCTGGCCTCCCAC
- Phenotype
-
MIM: 612547
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
WBSCR28
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1136647] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- WBSCR28
- Gene Name
- Williams-Beuren syndrome chromosome region 28
There are no transcripts associated with this gene.
View Full Product Details