Product Details

SNP ID
rs200077575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143222416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACCATTCCAATCTCTGGTTTGCTG[C/T]CTGGCTCAAAGTCTTCTATTGTCTT
Phenotype
MIM: 613964
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAS2R40 PubMed Links

Gene Details

Gene
TAS2R40
Gene Name
taste 2 receptor member 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_176882.1 338 Missense Mutation GCC,GTC A113V NP_795363.1

View Full Product Details