Product Details

SNP ID
rs200418776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:33097300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAACTGCTCTAACTTTTGGTTGC[C/T]TTTGATAAAGAAAGGGCATTCTTTG
Phenotype
MIM: 607331
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RP9 PubMed Links

Gene Details

Gene
RP9
Gene Name
retinitis pigmentosa 9 (autosomal dominant)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203288.1 870 Intron NP_976033.1
XM_011515468.2 870 Missense Mutation AGC,GGC S92G XP_011513770.1

View Full Product Details