Product Details

SNP ID
rs200344297
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143316332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTACGGACTGCCCTCTGAGAATGG[C/G]GGCCTCCAGCACAGGCTCCGGAAGG
Phenotype
MIM: 600639 MIM: 118425
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CASP2 PubMed Links

Gene Details

Gene
CASP2
Gene Name
caspase 2
There are no transcripts associated with this gene.

Gene
CLCN1
Gene Name
chloride voltage-gated channel 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000083.2 207 Silent Mutation GGC,GGG G40G NP_000074.2
XM_011515782.2 207 Intron XP_011514084.1
XM_017011739.1 207 Intron XP_016867228.1
XM_017011740.1 207 Intron XP_016867229.1

View Full Product Details