Product Details

SNP ID
rs200257708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:142939719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGAACGGGAGCCCAGGCCCCA[A/G]AGATGGGAGCCAGACAGAGAAAACG
Phenotype
MIM: 613883 MIM: 606679
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C7orf34 PubMed Links

Gene Details

Gene
C7orf34
Gene Name
chromosome 7 open reading frame 34
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178829.4 214 Missense Mutation AAA,AGA K58R NP_849151.2
Gene
KEL
Gene Name
Kell blood group, metallo-endopeptidase
There are no transcripts associated with this gene.

Gene
TRPV5
Gene Name
transient receptor potential cation channel subfamily V member 5
There are no transcripts associated with this gene.

View Full Product Details