Product Details

SNP ID
rs200881614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74220720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCGCAGGAAGCAGACACGGGTC[A/G]GAGGAAGCCTACATGTGAGTGACCT
Phenotype
MIM: 605719
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAT2 PubMed Links

Gene Details

Gene
LAT2
Gene Name
linker for activation of T-cells family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014146.3 708 Silent Mutation TCA,TCG S106S NP_054865.2
NM_032463.2 708 Silent Mutation TCA,TCG S106S NP_115852.1
NM_032464.2 708 Silent Mutation TCA,TCG S106S NP_115853.2
XM_011516558.2 708 Silent Mutation TCA,TCG S106S XP_011514860.1

View Full Product Details