Product Details
- SNP ID
-
rs200506099
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:5882494 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATAACCAATTCTCTGTTCTTCAGA[C/G]CCAGACACTTTTGAACCCCAAAAAT
- Phenotype
-
MIM: 164795
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
OCM
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150764259,rs35069590] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- OCM
- Gene Name
- oncomodulin
View Full Product Details