Product Details

SNP ID
rs202215700
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:81705466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCAGAGTCACCTTCCCTCGATGA[C/T]GCTGGCTGCATTTCTCATTTCCCAT
Phenotype
MIM: 142409
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HGF PubMed Links

Gene Details

Gene
HGF
Gene Name
hepatocyte growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000601.5 2161 Missense Mutation CAT,CGT H645R NP_000592.3
NM_001010931.2 2161 Intron NP_001010931.1
NM_001010932.2 2161 Missense Mutation CAT,CGT H640R NP_001010932.1
NM_001010933.2 2161 Intron NP_001010933.1
NM_001010934.2 2161 Intron NP_001010934.1
XM_006715956.2 2161 Missense Mutation CAT,CGT H645R XP_006716019.1
XM_011516115.2 2161 Missense Mutation CAT,CGT H640R XP_011514417.1
XM_017012097.1 2161 Intron XP_016867586.1
XM_017012098.1 2161 Intron XP_016867587.1

View Full Product Details