Product Details

SNP ID
rs201551577
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19144992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAAACGTGGGCACCTTCCTCCGC[A/G]GCTGGTCAAAGGCCTCGTTGAGGTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FERD3L PubMed Links

Gene Details

Gene
FERD3L
Gene Name
Fer3-like bHLH transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152898.2 430 Missense Mutation CCG,CTG P124L NP_690862.1

View Full Product Details