Product Details

SNP ID
rs201422718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149764885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCGGGCAGGAGAAGGGGCGGGA[A/G]CCTGTGTGGATCGCCTGGTGGCGGA
Phenotype
MIM: 614040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF467 PubMed Links

Gene Details

Gene
ZNF467
Gene Name
zinc finger protein 467
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005249959.4 1730 Silent Mutation XP_005250016.1
XM_005249960.4 1730 Silent Mutation XP_005250017.1
XM_005249961.4 1730 Silent Mutation XP_005250018.1
XM_006715864.3 1730 Silent Mutation XP_006715927.1
XM_011515855.2 1730 Silent Mutation XP_011514157.1
XM_011515856.2 1730 Silent Mutation XP_011514158.1
XM_011515857.2 1730 Silent Mutation XP_011514159.1
XM_011515858.1 1730 Silent Mutation XP_011514160.1
XM_017011799.1 1730 Silent Mutation XP_016867288.1

View Full Product Details