Product Details

SNP ID
rs201269559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:40134519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCGCTACCCCAACCTCCTCCAC[C/G]CGGACCAGGGTAAGGAGGAGTTGGG
Phenotype
MIM: 609188 MIM: 609187
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MPLKIP PubMed Links

Gene Details

Gene
MPLKIP
Gene Name
M-phase specific PLK1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138701.3 134 Missense Mutation CGT,GGT R17G NP_619646.1
Gene
SUGCT
Gene Name
succinyl-CoA:glutarate-CoA transferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193311.1 134 Intron NP_001180240.1
NM_001193312.1 134 Intron NP_001180241.1
NM_001193313.1 134 Intron NP_001180242.1
NM_024728.2 134 Intron NP_079004.1
XM_006715775.3 134 Intron XP_006715838.1
XM_011515525.2 134 Intron XP_011513827.1
XM_011515526.2 134 Intron XP_011513828.1
XM_011515527.2 134 Intron XP_011513829.1
XM_011515528.2 134 Intron XP_011513830.1
XM_011515529.2 134 Intron XP_011513831.1
XM_011515530.2 134 Intron XP_011513832.1
XM_017012621.1 134 Intron XP_016868110.1
XM_017012622.1 134 Intron XP_016868111.1

View Full Product Details