Product Details

SNP ID
rs201535763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:149764806 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGGGCCGCGTGGGCGGCTTCGC[C/T]GTGAATGAGCTGGTGCCGCACCAGG
Phenotype
MIM: 614040
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF467 PubMed Links

Gene Details

Gene
ZNF467
Gene Name
zinc finger protein 467
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005249959.4 1809 Missense Mutation CGC,TGC R566C XP_005250016.1
XM_005249960.4 1809 Missense Mutation CGC,TGC R566C XP_005250017.1
XM_005249961.4 1809 Missense Mutation CGC,TGC R566C XP_005250018.1
XM_006715864.3 1809 Missense Mutation CGC,TGC R607C XP_006715927.1
XM_011515855.2 1809 Missense Mutation CGC,TGC R817C XP_011514157.1
XM_011515856.2 1809 Missense Mutation CGC,TGC R817C XP_011514158.1
XM_011515857.2 1809 Missense Mutation CGC,TGC R780C XP_011514159.1
XM_011515858.1 1809 Missense Mutation CGC,TGC R566C XP_011514160.1
XM_017011799.1 1809 Missense Mutation CGC,TGC R741C XP_016867288.1

View Full Product Details